2-(Cyclopropyl)benzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (2-Cyclopropylphenyl)methanol

A233195-100MG A233195-250MG A233195-5G A233195-1G
103810-874EA 138 USD
103810-874 103850-634 76874-284 76874-286 77089-586 77303-958
2-(Cyclopropyl)benzyl alcohol 97%
2-(Cyclopropyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.202 g/mol
Boiling Pt: 264.3±9.0 °C (760 mm Hg)
Flash Pt: 122.8 °C
Storage Temperature: Ambient
MDL Number: MFCD20921818
CAS Number: 118184-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR