2-Chloro-3-thiopheneboronic acid 97%
Supplier: Ambeed
|
Warning
|
A221218-25G
A221218-5G
A221218-100MG
A221218-10G
A221218-250MG
A221218-1G
103818-906EA
27
USD
103818-906
103849-788
77088-388
77088-390
77275-484
77275-486
77275-488
77303-584
2-Chloro-3-thiopheneboronic acid 97%
2-Chloro-3-thiopheneboronic acid
Precaución: for Research Use Only
Formula:
C₄H₄BClO₂S Boiling Pt: 322.7 °C (760 mm Hg) Flash Pt: 149 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05662391 CAS Number: 177734-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...