(2-Chloro-4-pyridinyl)methanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol
A118211-100G
A118211-10G
A118211-250MG
A118211-1G
A118211-5G
A118211-25G
103799-006EA
14
USD
103799-006
103824-894
77009-274
77009-276
77009-278
77297-586
77311-686
77683-065
(2-Chloro-4-pyridinyl)methanol 98%
(2-Chloro-4-pyridinyl)methanol
Precaución: For research use only.
Formula:
C₆H₆ClNO MW: 143.57 g/mol Melting Pt: 59…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD06858767 CAS Number: 100704-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...