2-Chloro-4-pyridineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-Chloropyridin-4-yl)boronic acid, 2-Chloro-4-pyridinylboronic acid, 2-Chloropyridine-4-boronic acid

A305722-1G A305722-100G A305722-10G A305722-250MG A305722-25G A305722-5G
103814-448EA 10 USD
103814-448 103835-614 76838-320 77015-050 77015-052 77015-054 77305-918 77305-920
2-Chloro-4-pyridineboronic acid 97%
2-Chloro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BClNO₂
Boiling Pt: 355.1 °C (760 mm Hg)
Melting Pt: 155…163 °C
Flash Pt: 168.6 °C
Storage Temperature: Freezer
MDL Number: MFCD03788416
CAS Number: 458532-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR