(2-Chloropyridin-3-yl)acetonitrile 97%
Supplier: Ambeed
|
Warning
|
A160023-10G
A160023-1G
A160023-25G
A160023-5G
103805-316EA
15
USD
103805-316
103843-546
77028-588
77319-452
77319-454
77319-456
(2-Chloropyridin-3-yl)acetonitrile 97%
(2-Chloropyridin-3-yl)acetonitrile
Precaución: for Research Use Only
Formula:
C₇H₅ClN₂ MW: 152.5800 g/mol Boiling Pt: 294.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09923767 CAS Number: 101012-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...