2-Chlorophenylhydrazinium chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chlorophenylhydrazine hydrochloride, (2-Chlorophenyl)hydrazine hydrochloride, 1-(2-Chlorophenyl)hydrazine hydrochloride
A209556-25G
A209556-100G
A209556-10G
A209556-500G
A209556-5G
103848-634EA
6
USD
103848-634
77086-982
77112-416
77112-418
77112-420
77303-100
2-Chlorophenylhydrazinium chloride 97%
2-Chlorophenylhydrazinium chloride
Precaución: for Research Use Only
Formula:
C₆H₈Cl₂N₂ Boiling Pt: 252.1 °C (760 mm Hg) Melting Pt: 200…203 °C Flash Pt: 106.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012928 CAS Number: 41052-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...