(2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidin-6-yl)methanol 98%

Supplier: Ambeed
Warning

A247616-1G A247616-25G A247616-5G
103811-784EA 28 USD
103811-784 103851-620 77304-418 77304-420 77304-422
(2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidin-6-yl)methanol 98%
(2-Chloro-7-cyclopentyl-7H-pyrrolo[2,3-d]pyrimidin-6-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClN₃O
MW: 251.71 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29072793
CAS Number: 1374639-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR