(2-Chloro-6-nitrophenyl)methanol 97%

Supplier: Ambeed
Warning

A106076-10G A106076-250MG A106076-1G A106076-5G
103796-994EA 33 USD
103796-994 103822-746 77135-706 77135-708 77309-608 77309-610
(2-Chloro-6-nitrophenyl)methanol 97%
(2-Chloro-6-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00086565
CAS Number: 50907-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR