(2-Chloro-4-methylphenyl)methanamine 96%

Supplier: Ambeed
Danger

A554131-100MG A554131-250MG A554131-1G
103828-702EA 94 USD
103828-702 76819-076 76819-078 76843-916
(2-Chloro-4-methylphenyl)methanamine 96%
(2-Chloro-4-methylphenyl)methanamine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
Storage Temperature: Refrigerator
MDL Number: MFCD06213616
CAS Number: 69957-96-6
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR