(2-Chloro-4-isopropoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A176441-1G A176441-250MG A176441-5G
103807-136EA 68 USD
103807-136 103845-464 77082-820 77096-638 77301-448
(2-Chloro-4-isopropoxyphenyl)boronic acid 98%
(2-Chloro-4-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₃
Boiling Pt: 356 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02684319
CAS Number: 313545-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR