2-Chloro-(4-methoxycarbonyl)phenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4-(methoxycarbonyl)phenylboronic acid, 2-Chloro-4-(methoxycarbonyl)benzeneboronic acid, (2-Chloro-4-(methoxycarbonyl)phenyl)boronic acid
A203645-10G
A203645-250MG
A203645-25G
A203645-5G
103817-474EA
16
USD
103817-474
103848-134
76975-298
76975-300
77086-262
77086-264
2-Chloro-(4-methoxycarbonyl)phenylboronic acid 97%
2-Chloro-(4-methoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₈BClO₄ MW: 214.4107 g/mol Boiling Pt: 386.6 °C (760 mm Hg) Melting Pt: 140…142 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08689530 CAS Number: 603122-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...