2-Chloro-3-fluoro-hydrazine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-3-fluorophenyl)hydrazine hydrochloride

A354398-1G A354398-25G A354398-5G A354398-10G
103826-122EA 7 USD
103826-122 103837-126 76840-244 76840-246 76840-248 77155-032
2-Chloro-3-fluoro-hydrazine hydrochloride 98%
2-Chloro-3-fluoro-hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₆ClFN₂·ClH
MW: 197.038 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18393091
CAS Number: 1138036-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR