2-Chloro-3-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-3-fluorobenzeneboronic acid, (2-Chloro-3-fluorophenyl)boronic acid

A207500-100MG A207500-10G A207500-250MG A207500-1G A207500-5G
103817-798EA 15 USD
103817-798 103848-480 77030-356 77030-358 77030-360 77086-754 77086-756
2-Chloro-3-fluorophenylboronic acid 97%
2-Chloro-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
Boiling Pt: 313.4 °C (760 mm Hg)
Melting Pt: 80…84 °C
Flash Pt: 143.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363741
CAS Number: 871329-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR