β-Bromostyrene (mixture of cis and trans isomers) 95%

Supplier: Ambeed
Warning

Synonyms: (2-Bromovinyl)benzene, β-Bromostyrene

A117091-5G A117091-25G A117091-100G
103824-698EA 12 USD
103824-698 77311-486 77549-344 77623-986
β-Bromostyrene (mixture of cis and trans isomers) 95%
β-Bromostyrene (mixture of cis and trans isomers)

Precaución: for Research Use Only

Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 110…112 °C (760 mm Hg)
Melting Pt: 7 °C
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000185
CAS Number: 103-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR