(2-Bromopyridin-4-yl)methanamine 97%

Supplier: Ambeed
Warning

A150324-100MG A150324-5G
103804-144EA 142 USD
103804-144 103842-280 76984-602 77317-790
(2-Bromopyridin-4-yl)methanamine 97%
(2-Bromopyridin-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Boiling Pt: 292.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07367880
CAS Number: 858362-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR