2-Bromo-4-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Bromopyridin-4-yl)boronic acid, 2-Bromo-4-pyridinylboronic acid, 2-Bromopyridine-4-boronic acid

A130956-10G A130956-250MG A130956-1G
103801-502EA 40 USD
103801-502 77258-442 77258-444 77298-436
2-Bromo-4-pyridineboronic acid 98%
2-Bromo-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BBrNO₂
MW: 201.81 g/mol
Boiling Pt: 375.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06798244
CAS Number: 458532-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR