2-Bromobenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (2-Bromophenyl)methanamine

A101201-10G A101201-25G A101201-1G A101201-5G
103795-854EA 6 USD
103795-854 103821-538 77107-202 77107-204 77282-676 77282-678
2-Bromobenzylamine 98%
2-Bromobenzylamine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 243.5 °C (760 mm Hg)
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025572
CAS Number: 3959-05-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR