2-Bromobenzophenone 95%

Supplier: Ambeed
Warning

A451241-100G A451241-10G A451241-1G A451241-25G A451241-5G
103827-790EA 14 USD
103827-790 103838-860 76725-572 76725-574 76725-576 76842-680 76842-682 76842-684
2-Bromobenzophenone 95%
2-Bromobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉BrO
Melting Pt: 41…42 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00032462
CAS Number: 13047-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR