Phenethyl bromide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Bromoethyl)benzene, 2-Phenylethyl bromide
A136962-100G
A136962-25G
A136962-500G
A136962-1000G
A136962-10G
103808-546EA
63
USD
103808-546
77137-136
77137-138
77137-140
77315-316
77566-722
Phenethyl bromide 98%
Phenethyl bromide
Precaución: For research use only.
Formula:
BrCH₂CH₂C₆H₅ MW: 185.06 g/mol Boiling Pt: 220…221 °C (760 mm Hg) Melting Pt: …67.5 °C Flash Pt: 96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000240 CAS Number: 103-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...