2-Bromo-6-nitrobenzylalkohol 97%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-6-nitrophenyl)methanol

A113037-100MG A113037-250MG A113037-1G
103798-150EA 19 USD
103798-150 103823-986 76810-030 76810-032 77310-822
2-Bromo-6-nitrobenzylalkohol 97%
2-Bromo-6-nitrobenzylalkohol

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
Boiling Pt: 332.6 °C (760 mm Hg)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD11036298
CAS Number: 861106-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR