2-Bromo-5-fluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-5-fluorophenyl)methanol

A456139-1G A456139-5G A456139-100G A456139-10G A456139-500G A456139-25G
103827-854EA 5 USD
103827-854 103838-928 76842-776 76842-778 77004-354 77004-356 77004-358 77307-396
2-Bromo-5-fluorobenzyl alcohol 98%
2-Bromo-5-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 252.5 °C (760 mm Hg)
Melting Pt: 91…94 °C
Flash Pt: 106.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00142876
CAS Number: 202865-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR