2-Bromo-5-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-5-fluorophenyl)boronic acid

A590649-250MG A590649-5G
103828-926EA 51 USD
103828-926 77280-214 77749-214
2-Bromo-5-fluorophenylboronic acid 98%
2-Bromo-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
Boiling Pt: 326.9 °C (760 mm Hg)
Flash Pt: 151.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690232
CAS Number: 959996-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR