2-Bromo-5-chlorobenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-5-chlorophenyl)methanol

A170925-10G A170925-1G A170925-5G
103806-556EA 5 USD
103806-556 103844-832 76784-226 77081-952 77081-954
2-Bromo-5-chlorobenzyl alcohol 97%
2-Bromo-5-chlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.4800 g/mol
Boiling Pt: 290.1 °C (760 mm Hg)
Melting Pt: 81…83 °C
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD09261147
CAS Number: 60666-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR