2-Bromo-3-iodobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-3-iodophenyl)methanol

A263866-250MG A263866-1G A263866-5G
103812-644EA 222 USD
103812-644 103852-536 76874-936 77092-180 77092-182
2-Bromo-3-iodobenzyl alcohol 98%
2-Bromo-3-iodobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrIO
MW: 312.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520410
CAS Number: 1261644-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR