2-Aminopyridine-4-methanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-pyridinylmethanol, (2-Aminopyridin-4-yl)-methanol, (2-Aminopyridin-4-yl)methanol, (2-Aminopyrid-4-yl)methanol

A187483-100G A187483-10G A187483-1G A187483-25G A187483-5G
103816-018EA 7 USD
103816-018 103846-608 77012-174 77012-176 77012-178 77084-298 77084-300 77084-302
2-Aminopyridine-4-methanol 97%
2-Aminopyridine-4-methanol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
Melting Pt: 80…84 °C
Flash Pt: 153.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03791261
CAS Number: 105250-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR