(2-Aminopyridin-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

A207117-100MG A207117-250MG A207117-1G
103817-762EA 114 USD
103817-762 103848-442 77030-326 77030-328 77086-714
(2-Aminopyridin-4-yl)boronic acid 97%
(2-Aminopyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₇BN₂O₂
MW: 137.93 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11520501
CAS Number: 903513-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR