2-Aminobenzyl alcohol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol
A102134-100G
A102134-10G
A102134-25G
A102134-500G
A102134-5G
A102134-1000G
103822-004EA
5
USD
103822-004
77093-548
77093-550
77093-552
77093-554
77308-880
77684-930
2-Aminobenzyl alcohol 98%
2-Aminobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₉NO Boiling Pt: 275 °C (760 mm Hg) Melting Pt: 81…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007749 CAS Number: 5344-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...