2-Aminophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminobenzeneboronic acid, (2-Aminophenyl)boronic acid, (2-Aminophenyl)metaboric acid, Dihydroxy(2-aminophenyl)borane

A100028-100G A100028-100MG A100028-250MG A100028-25G A100028-1G A100028-5G A100028-10G
103795-208EA 8 USD
103795-208 103820-930 76809-182 76809-184 76809-186 76809-188 77296-546 77296-548 77548-852
2-Aminophenylboronic acid 97%
2-Aminophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₂
MW: 136.95 g/mol
Boiling Pt: 346.9 °C (760 mm Hg)
Melting Pt: 188…190 °C
Flash Pt: 163.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074645
CAS Number: 5570-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR