(2-Aminoethyl)phosphonic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-AEP, 2-Aminoethylphosphonic acid

A167971-1G A167971-250MG
103806-232EA 63 USD
103806-232 103844-490 77081-524 77259-824
(2-Aminoethyl)phosphonic acid 97%
(2-Aminoethyl)phosphonic acid

Precaución: for Research Use Only

Formula: C₂H₈NO₃P
MW: 125.064 g/mol
Boiling Pt: 352.5 °C (760 mm Hg)
Melting Pt: 296 °C
Storage Temperature: Ambient
MDL Number: MFCD00008182
CAS Number: 2041-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR