(2-Amino-5-ethylthiophen-3-yl)(2-chlorophenyl)methanone 97%

Supplier: Ambeed
Warning

A439170-1G A439170-25G A439170-100G A439170-250MG
103827-642EA 6 USD
103827-642 103838-708 76842-458 76842-460 77102-396 77102-398
(2-Amino-5-ethylthiophen-3-yl)(2-chlorophenyl)methanone 97%
(2-Amino-5-ethylthiophen-3-yl)(2-chlorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNOS
Boiling Pt: 454.8 °C (760 mm Hg)
Flash Pt: 228.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04107940
CAS Number: 50508-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR