2-Amino-2',5-dichlorobenzophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Amino-5-chlorophenyl)(2-chlorophenyl)methanone
A185784-100G
A185784-25G
A185784-5G
A185784-1G
103815-884EA
5
USD
103815-884
103846-454
76813-736
77084-088
77084-090
77301-994
2-Amino-2',5-dichlorobenzophenone 98%
2-Amino-2',5-dichlorobenzophenone
Precaución: for Research Use Only
Formula:
C₁₃H₉Cl₂NO MW: 266.13 g/mol Boiling Pt: 453.6 °C (760 mm Hg) Melting Pt: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007840 CAS Number: 2958-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...