2-Amino-5-bromobenzylalkohol 98%
Supplier: Ambeed
|
Warning
|
A194142-1G
A194142-5G
A194142-250MG
A194142-10G
103816-622EA
10
USD
103816-622
103847-232
77085-146
77085-148
77139-178
77685-475
2-Amino-5-bromobenzylalkohol 98%
2-Amino-5-bromobenzylalkohol
Precaución: for Research Use Only
Formula:
C₇H₈BrNO Boiling Pt: 329.6 °C (760 mm Hg) Melting Pt: 111…112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08234509 CAS Number: 20712-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...