(2-Amino-5-(trifluoromethyl)phenyl)methanol 97%

Supplier: Ambeed
Warning

A301949-100MG A301949-1G
103814-320EA 108 USD
103814-320 103835-476 76787-762 77305-844
(2-Amino-5-(trifluoromethyl)phenyl)methanol 97%
(2-Amino-5-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11849935
CAS Number: 220107-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR