(2-Amino-4-chlorophenyl)methanol 97%

Supplier: Ambeed
Warning

A296262-1G A296262-5G A296262-10G A296262-250MG
103814-086EA 46 USD
103814-086 103835-226 76837-890 76837-892 76988-708 76988-710
(2-Amino-4-chlorophenyl)methanol 97%
(2-Amino-4-chlorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD04037367
CAS Number: 37585-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR