2-Amino-3-methylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (2-Amino-3-methylphenyl)methanol

A264536-100MG A264536-1G
103812-684EA 35 USD
103812-684 103852-576 77250-214 77304-898
2-Amino-3-methylbenzyl alcohol 95%
2-Amino-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.179 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00075027
CAS Number: 57772-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR