(2-Amino-3,5-dibromophenyl)methanol 95%

Supplier: Ambeed
Warning

A243756-250MG A243756-1G
103811-548EA 37 USD
103811-548 103851-372 76976-542 77090-576
(2-Amino-3,5-dibromophenyl)methanol 95%
(2-Amino-3,5-dibromophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₇Br₂NO
MW: 280.95 g/mol
Boiling Pt: 366.3 °C (760 mm Hg)
Melting Pt: 146…148 °C
Flash Pt: 175.3 °C
Storage Temperature: Ambient
MDL Number: MFCD06202693
CAS Number: 50739-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR