(2-Amino-2,3-dihydro-1H-inden-2-yl)phosphonic acid hydrochloride 97%

Supplier: Ambeed
Warning

A110664-100MG A110664-250MG A110664-1G
103797-702EA 374 USD
103797-702 103823-510 76821-780 76821-782 77310-316
(2-Amino-2,3-dihydro-1H-inden-2-yl)phosphonic acid hydrochloride 97%
(2-Amino-2,3-dihydro-1H-inden-2-yl)phosphonic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClNO₃P
Storage Temperature: Ambient
MDL Number: MFCD23135861
CAS Number: 1416354-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR