2,6-Dimethyl-4-pyridineboronic acid 97%

Supplier: Ambeed
Warning

A137034-100MG A137034-250MG A137034-1G A137034-5G
103802-392EA 32 USD
103802-392 76811-570 76811-572 77315-330 77315-332
2,6-Dimethyl-4-pyridineboronic acid 97%
2,6-Dimethyl-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₂
Boiling Pt: 252 °C (760 mm Hg)
Flash Pt: 106 °C
Storage Temperature: Freezer
MDL Number: MFCD11042425
CAS Number: 846548-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR