2,5-Difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2,5-Difluorophenyl)boronic acid, 2,5-Difluorobenzeneboronic acid, (2,5-Difluorophenyl)metaboric acid, Dihydroxy(2,5-difluorophenyl)borane

A169453-25G A169453-5G A169453-100G A169453-10G A169453-1G A169453-500G
103844-646EA 14 USD
103844-646 77081-730 77081-732 77286-752 77286-754 77286-756 77615-416
2,5-Difluorophenylboronic acid 98%
2,5-Difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Boiling Pt: 271.3 °C (760 mm Hg)
Melting Pt: 105…110 °C
Flash Pt: 117.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01863171
CAS Number: 193353-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR