2,5-Dichlorophenylhydrazinium chloride 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Dichlorophenylhydrazine hydrochloride, 1-(2,5-Dichlorophenyl)hydrazine hydrochloride, (2,5-Dichlorophenyl)hydrazine hydrochloride

A819211-25G A819211-5G
103841-122EA 11 USD
103841-122 76845-362 76845-364
2,5-Dichlorophenylhydrazinium chloride 98%
2,5-Dichlorophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Boiling Pt: 266.8 °C (760 mm Hg)
Flash Pt: 115.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00052266
CAS Number: 50709-35-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR