2,4-Difluorobenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (2,4-Difluorophenyl)methanamine

A149785-10G A149785-1G A149785-25G A149785-5G
103804-080EA 5 USD
103804-080 103842-208 76768-118 77317-718 77317-720 77317-722
2,4-Difluorobenzylamine 98%
2,4-Difluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010142
CAS Number: 72235-52-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR