2,4-Difluorophenylhydrazinium chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4-Difluorophenylhydrazine hydrochloride, 1-(2,4-Difluorophenyl)hydrazine hydrochloride
A322750-5G
A322750-100G
A322750-10G
A322750-500G
A322750-25G
103836-244EA
7
USD
103836-244
76839-164
77290-318
77290-320
77290-322
77306-272
2,4-Difluorophenylhydrazinium chloride 97%
2,4-Difluorophenylhydrazinium chloride
Precaución: for Research Use Only
Formula:
C₆H₇ClF₂N₂ Melting Pt: 210 °C Flash Pt: 65.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013383 CAS Number: 51523-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...