2,4-Dichlorophenylhydrazinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: (2,4-Dichlorophenyl)hydrazine hydrochloride, 2,4-Dichlorophenylhydrazine hydrochloride, 1-(2,4-Dichlorophenyl)hydrazine hydrochloride

A152259-100G A152259-10G A152259-5G A152259-25G A152259-500G
103842-534EA 6 USD
103842-534 77286-176 77286-178 77299-938 77318-148 77615-236
2,4-Dichlorophenylhydrazinium chloride 98%
2,4-Dichlorophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Boiling Pt: 268.8 °C (760 mm Hg)
Melting Pt: 220…224 °C
Flash Pt: 116.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00012929
CAS Number: 5446-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR