(2,4,6-Trimethoxyphenyl)methanethiol 98%

Supplier: Ambeed
Danger

A296009-1G A296009-5G A296009-250MG
103814-082EA 59 USD
103814-082 103835-216 76837-878 76837-880 76875-582
(2,4,6-Trimethoxyphenyl)methanethiol 98%
(2,4,6-Trimethoxyphenyl)methanethiol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃S
Boiling Pt: 326 °C (760 mm Hg)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD14586337
CAS Number: 212555-23-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR