2,4,6-Triisopropylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,4,6-Triisopropylbenzeneboronic acid, (2,4,6-Triisopropylphenyl)boronic acid

A232829-10G A232829-250MG A232829-1G A232829-25G A232829-5G
103810-856EA 6 USD
103810-856 103850-610 76826-520 76826-522 77089-544 77089-546 77089-548
2,4,6-Triisopropylphenylboronic acid 98%
2,4,6-Triisopropylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₅BO₂
MW: 248.16 g/mol
Boiling Pt: 346.8 °C (760 mm Hg)
Melting Pt: 165 °C
Flash Pt: 163.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683099
CAS Number: 154549-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR