2,3-Dimethylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (2,3-Dimethylphenyl)methanol

A234784-1G A234784-5G
103810-976EA 14 USD
103810-976 103850-732 77303-994 77303-996
2,3-Dimethylbenzyl alcohol 95%
2,3-Dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 225.9 °C (760 mm Hg)
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00046742
CAS Number: 13651-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR