2,3-Dimethyl-2,3-diphenylbutane 95%

Supplier: Ambeed
Warning

Synonyms: 1,1'-(1,1,2,2-Tetramethylethylene)dibenzene, (2,3-Dimethylbutane-2,3-diyl)dibenzene

A271443-25G A271443-100G A271443-10G A271443-500G A271443-1000G
103809-890EA 271 USD
103809-890 77092-736 77250-366 77250-368 77250-370 77305-078
2,3-Dimethyl-2,3-diphenylbutane 95%
2,3-Dimethyl-2,3-diphenylbutane

Precaución: for Research Use Only

Formula: H₃CC(CH₃)(C₆H₅)C(CH₃)(C₆H₅)CH₃
MW: 238.37 g/mol
Boiling Pt: 306.1 °C (760 mm Hg)
Melting Pt: 90…110 °C
Flash Pt: 142.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053713
CAS Number: 1889-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR