(2,3-Dimethyl-1H-indol-5-yl)methanamine 97%
Supplier: Ambeed
|
Warning
|
A329244-100MG
A329244-250MG
A329244-1G
103825-520EA
380
USD
103825-520
77003-004
77003-006
77306-394
(2,3-Dimethyl-1H-indol-5-yl)methanamine 97%
(2,3-Dimethyl-1H-indol-5-yl)methanamine
Precaución: for Research Use Only
Formula:
C₁₁H₁₄N₂ MW: 174.24 g/mol Melting Pt: 153…154 °C at N/A mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD00509410 CAS Number: 5054-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...