2,3-Dihydro-1-benzofuran-7-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2,3-Dihydrobenzofuran-7-yl)boronic acid

A273368-250MG A273368-5G A273368-1G
103813-140EA 33 USD
103813-140 103853-060 76875-130 77092-858 77305-128
2,3-Dihydro-1-benzofuran-7-boronic acid 98%
2,3-Dihydro-1-benzofuran-7-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
Boiling Pt: 149 °C (760 mm Hg)
Flash Pt: 105 °C
Storage Temperature: Freezer
MDL Number: MFCD06801700
CAS Number: 685514-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR