(2,3-Dihydrobenzofuran-6-yl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A221632-100MG
A221632-250MG
A221632-1G
103818-938EA
138
USD
103818-938
103849-816
77013-270
77013-272
77088-416
(2,3-Dihydrobenzofuran-6-yl)boronic acid 98%
(2,3-Dihydrobenzofuran-6-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BO₃ Boiling Pt: 160…163 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD22398271 CAS Number: 763120-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...